Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-benzylpiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
50541-93-0
C12H18N2
190.29
MFCD00006504
YUBDLZGUSSWQSS-UHFFFAOYSA-N
4-amino-1-benzylpiperidine,1-benzyl-4-aminopiperidine,1-benzyl-piperidin-4-ylamine,n-benzyl-4-amino piperidine,1-benzyl-4-piperidylamine,4-piperidinamine, 1-phenylmethyl,4-amino-1-n-benzylpiperidine,4-amino-n-benzylpiperidine,1-phenylmethyl-4-piperidinamine,n-benzyl-4-aminopiperidine
415852
1-benzylpiperidin-4-amine
C1CN(CCC1N)CC2=CC=CC=C2
50541-93-0
C12H18N2
190.29
MFCD00006504
YUBDLZGUSSWQSS-UHFFFAOYSA-N
4-amino-1-benzylpiperidine,1-benzyl-4-aminopiperidine,1-benzyl-piperidin-4-ylamine,n-benzyl-4-amino piperidine,1-benzyl-4-piperidylamine,4-piperidinamine, 1-phenylmethyl,4-amino-1-n-benzylpiperidine,4-amino-n-benzylpiperidine,1-phenylmethyl-4-piperidinamine,n-benzyl-4-aminopiperidine
415852
1-benzylpiperidin-4-amine
C1CN(CCC1N)CC2=CC=CC=C2
Packaging Glass bottle
Quantity 5g
Specific Gravity 0.93
Flash Point >110°C
Density 0.9300g/mL
Boiling Point 152°C (9.7mmHg)
Refractive Index 1.5410 to 1.543
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ether
Formula Weight 190.29
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Amino-1-benzylpiperidine, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 256-620-9