Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-1-butanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13325-10-5
C4H11NO
89.14
MFCD00008230
BLFRQYKZFKYQLO-UHFFFAOYSA-N
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
25868
4-aminobutan-1-ol
C(CCO)CN
13325-10-5
C4H11NO
89.14
MFCD00008230
BLFRQYKZFKYQLO-UHFFFAOYSA-N
4-amino-1-butanol, 4-aminobutanol, 1-butanol, 4-amino, 4-amino-butan-1-ol, 4-hdyroxybutylamine, 4-hydroxybutylamine, 4-hydroxy-n-butylamine, butanolamine, 4-amino butanol, 4-amino-butanol
25868
4-aminobutan-1-ol
C(CCO)CN
Specific Gravity 0.96
Infrared Spectrum Authentic
Color Colorless to Yellow
Density 0.9600g/mL
Melting Point 16.0°C to 18.0°C
Refractive Index 1.4605 to 1.4625
Flash Point 107°C
Boiling Point 206.0°C
Packaging Glass bottle
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula H2N(CH2)4OH
Beilstein 04, 291
Solubility Information Solubility in water: miscible
Formula Weight 89.14
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Amino-1-butanol, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 236-364-4