Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2,2,6,6-tetramethylpiperidine, 98%

Click to view available options
Quantity:
1g
5g
25g
36768-62-4
C9H20N2
156.273
MFCD00005984
FTVFPPFZRRKJIH-UHFFFAOYSA-N
4-amino-2,2,6,6-tetramethylpiperidine,triacetonediamine,2,2,6,6-tetramethyl-4-aminopiperidine,4-piperidinamine, 2,2,6,6-tetramethyl,unii-etf220q65r,2,2,6,6-tetramethyl-4-piperidylamine,4-amino-2,2,6,6-tetramethyl-piperidine,piperidine, 4-amino-2,2,6,6-tetramethyl,2,2,6,6-tetramethyl-4-piperidinamine,4-amino-2,2,6,6-tetramethyl piperidine
37524
2,2,6,6-tetramethylpiperidin-4-amine
CC1(CC(CC(N1)(C)C)N)C
36768-62-4
C9H20N2
156.273
MFCD00005984
FTVFPPFZRRKJIH-UHFFFAOYSA-N
4-amino-2,2,6,6-tetramethylpiperidine,triacetonediamine,2,2,6,6-tetramethyl-4-aminopiperidine,4-piperidinamine, 2,2,6,6-tetramethyl,unii-etf220q65r,2,2,6,6-tetramethyl-4-piperidylamine,4-amino-2,2,6,6-tetramethyl-piperidine,piperidine, 4-amino-2,2,6,6-tetramethyl,2,2,6,6-tetramethyl-4-piperidinamine,4-amino-2,2,6,6-tetramethyl piperidine
37524
2,2,6,6-tetramethylpiperidin-4-amine
CC1(CC(CC(N1)(C)C)N)C
Flash Point 75°C (167°F)
Density 0.912
Quantity 1g
Boiling Point 188°C to 189°C
Melting Point 16°C to 18°C
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidine

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

EINECSNumber : 253-197-2

TSCA : Yes

Recommended Storage : Ambient temperatures