Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2,2,6,6-tetramethylpiperidine, 98%

Click to view available options
Quantity:
1g
5g
25g
36768-62-4
C9H20N2
156.273
MFCD00005984
FTVFPPFZRRKJIH-UHFFFAOYSA-N
4-amino-2,2,6,6-tetramethylpiperidine,triacetonediamine,2,2,6,6-tetramethyl-4-aminopiperidine,4-piperidinamine, 2,2,6,6-tetramethyl,unii-etf220q65r,2,2,6,6-tetramethyl-4-piperidylamine,4-amino-2,2,6,6-tetramethyl-piperidine,piperidine, 4-amino-2,2,6,6-tetramethyl,2,2,6,6-tetramethyl-4-piperidinamine,4-amino-2,2,6,6-tetramethyl piperidine
37524
2,2,6,6-tetramethylpiperidin-4-amine
CC1(CC(CC(N1)(C)C)N)C
36768-62-4
C9H20N2
156.273
MFCD00005984
FTVFPPFZRRKJIH-UHFFFAOYSA-N
4-amino-2,2,6,6-tetramethylpiperidine,triacetonediamine,2,2,6,6-tetramethyl-4-aminopiperidine,4-piperidinamine, 2,2,6,6-tetramethyl,unii-etf220q65r,2,2,6,6-tetramethyl-4-piperidylamine,4-amino-2,2,6,6-tetramethyl-piperidine,piperidine, 4-amino-2,2,6,6-tetramethyl,2,2,6,6-tetramethyl-4-piperidinamine,4-amino-2,2,6,6-tetramethyl piperidine
37524
2,2,6,6-tetramethylpiperidin-4-amine
CC1(CC(CC(N1)(C)C)N)C
Melting Point 16°C to 18°C
Flash Point 75°C (167°F)
Boiling Point 188°C to 189°C
Quantity 25g
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 4-Amino-2,2,6,6-tetramethylpiperidine

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA