Alfa Aesar™ 4-Amino-2,2-difluoro-1,3-benzodioxole, 97+%

Manufacturer: Alfa Aesar™ B2279703

Quantity 1g

 View more versions of this product

Catalog No. AAB2279703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106876-54-4
    Molecular Formula C7H5F2NO2
    Formula Weight 173.12
    MDL Number MFCD00792416
    UN Number UN2810
    Chemical Name or Material 4-Amino-2,2-difluoro-1,3-benzodioxole

    Specifications

    Formula Weight 173.12
    Quantity 1g
    MDL Number MFCD00792416
    UN Number UN2810
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C7H5F2NO2
    CAS 106876-54-4
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Amino-2,2-difluoro-1,3-benzodioxole
    Assay ≥97%

    Description

    Applications
    4-Amino-2,2-difluoro-1,3-benzodioxole is used in organic synthesis.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H312-H332-H315-H319
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit