Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2,3,5,6-tetrafluorobenzamide 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1548-74-9
C7H4F4N2O
208.116
MFCD00008000
CAERPAFTLPIDJT-UHFFFAOYSA-N
4-aminotetrafluorobenzamide,acmc-1cujp,4-amino-2,3,5,6-tetrafluorobenzamid,2,3,5,6-tetrafluoro-4-aminobenzamide,benzamide,4-amino-2,3,5,6-tetrafluoro,benzamide, 4-amino-2,3,5,6-tetrafluoro
73769
4-amino-2,3,5,6-tetrafluorobenzamide
C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)N
1548-74-9
C7H4F4N2O
208.116
MFCD00008000
CAERPAFTLPIDJT-UHFFFAOYSA-N
4-aminotetrafluorobenzamide,acmc-1cujp,4-amino-2,3,5,6-tetrafluorobenzamid,2,3,5,6-tetrafluoro-4-aminobenzamide,benzamide,4-amino-2,3,5,6-tetrafluoro,benzamide, 4-amino-2,3,5,6-tetrafluoro
73769
4-amino-2,3,5,6-tetrafluorobenzamide
C1(=C(C(=C(C(=C1F)F)N)F)F)C(=O)N
Quantity 5g
Color Beige
Packaging Glass bottle
Melting Point 183.0°C to 187.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2NC6F4CONH2
Formula Weight 208.11
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluorobenzamide, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.