Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2,3,5,6-tetrafluoropyridine, 98%

Click to view available options
Quantity:
1g
5g
1682-20-8
C5H2F4N2
166.079
MFCD00010855
PLVFNMQHMRRBAA-UHFFFAOYSA-N
4-amino-2,3,5,6-tetrafluoropyridine,4-aminotetrafluoropyridine,tetrafluoropyridin-4-amine,2,3,5,6-tetrafluoro-4-pyridinamine,2,3,5,6-tetrafluoro-4-pyridylamine,pubchem2992,acmc-1bzbr,4-amino-tetrafluoropyridine,4-amino-2,3,5,6-tetrafluorropyridine,4-pyridinamine, 2,3,5,6-tetrafluoro
1270911
2,3,5,6-tetrafluoropyridin-4-amine
C1(=C(C(=NC(=C1F)F)F)F)N
1682-20-8
C5H2F4N2
166.079
MFCD00010855
PLVFNMQHMRRBAA-UHFFFAOYSA-N
4-amino-2,3,5,6-tetrafluoropyridine,4-aminotetrafluoropyridine,tetrafluoropyridin-4-amine,2,3,5,6-tetrafluoro-4-pyridinamine,2,3,5,6-tetrafluoro-4-pyridylamine,pubchem2992,acmc-1bzbr,4-amino-tetrafluoropyridine,4-amino-2,3,5,6-tetrafluorropyridine,4-pyridinamine, 2,3,5,6-tetrafluoro
1270911
2,3,5,6-tetrafluoropyridin-4-amine
C1(=C(C(=NC(=C1F)F)F)F)N
Melting Point 84°C to 87°C
Quantity 1g
Beilstein 475870
Formula Weight 166.08
Percent Purity 98%
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluoropyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger