Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide, 98%

Click to view available options
Quantity:
25g
100g
52298-44-9
C14H16N2O4S
308.352
MFCD00025753
NFYQAEPHDGXJSY-UHFFFAOYSA-N
4-amino-2,5-dimethoxy-n-phenylbenzenesulphonamide,unii-54s41p93bi,2,5-dimethoxyaniline-4-sulfonanilide,benzenesulfonamide, 4-amino-2,5-dimethoxy-n-phenyl,2,5-dimethoxyaniline-4-sulfoanilide,acmc-1akg1,4-amino-2,5-dimethoxy-n-phenylbenzolsulfonamid,dsstox_cid_24686,dsstox_rid_80397,dsstox_gsid_44686
104143
4-amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
COC1=CC(=C(C=C1N)OC)S(=O)(=O)NC2=CC=CC=C2
52298-44-9
C14H16N2O4S
308.352
MFCD00025753
NFYQAEPHDGXJSY-UHFFFAOYSA-N
4-amino-2,5-dimethoxy-n-phenylbenzenesulphonamide,unii-54s41p93bi,2,5-dimethoxyaniline-4-sulfonanilide,benzenesulfonamide, 4-amino-2,5-dimethoxy-n-phenyl,2,5-dimethoxyaniline-4-sulfoanilide,acmc-1akg1,4-amino-2,5-dimethoxy-n-phenylbenzolsulfonamid,dsstox_cid_24686,dsstox_rid_80397,dsstox_gsid_44686
104143
4-amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
COC1=CC(=C(C=C1N)OC)S(=O)(=O)NC2=CC=CC=C2
Quantity 25g
Melting Point 184°C to 188°C
Beilstein 13660911
Formula Weight 308.35
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide

TSCA : TSCA

storageNote1 : -30° to -10°C