Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4'-Amino-2',6'-dibromoacetanilide, 99%

Click to view available options
Quantity:
5g
25g
336624-08-9
C8H8Br2N2O
307.973
MFCD02683078
IVRWBDFGHCBGBF-UHFFFAOYSA-N
n-4-amino-2,6-dibromophenyl acetamide,4'-amino-2',6'-dibromoacetanilide
24212084
N-(4-amino-2,6-dibromophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1Br)N)Br
336624-08-9
C8H8Br2N2O
307.973
MFCD02683078
IVRWBDFGHCBGBF-UHFFFAOYSA-N
n-4-amino-2,6-dibromophenyl acetamide,4'-amino-2',6'-dibromoacetanilide
24212084
N-(4-amino-2,6-dibromophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1Br)N)Br
Quantity 25g
Melting Point 246°C to 249°C
Formula Weight 307.97
Percent Purity 99%
Chemical Name or Material 4'-Amino-2',6'-dibromoacetanilide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning