Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2-chloro-5-nitropyrimidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
1920-66-7
C4H3ClN4O2
174.544
MFCD00127771
RZGOEIWDMVQJBQ-UHFFFAOYSA-N
4-amino-2-chloro-5-nitropyrimidine,4-pyrimidinamine, 2-chloro-5-nitro,2-chloro-4-amino-5-nitropyrimidine,2-chloro-5-nitro-pyrimidin-4-ylamine,2-chloro-5-nitro-4-pyrimidinamine,4-pyrimidinamine,2-chloro-5-nitro,acmc-209x0o,4-amino-5-nitro-2-chloropyrimidine,2-chloro-5-nitropyrimidine-4-amine,4-amino-2-chloro-5-nitropyrimidine
74716
2-chloro-5-nitropyrimidin-4-amine
C1=C(C(=NC(=N1)Cl)N)[N+](=O)[O-]
1920-66-7
C4H3ClN4O2
174.544
MFCD00127771
RZGOEIWDMVQJBQ-UHFFFAOYSA-N
4-amino-2-chloro-5-nitropyrimidine,4-pyrimidinamine, 2-chloro-5-nitro,2-chloro-4-amino-5-nitropyrimidine,2-chloro-5-nitro-pyrimidin-4-ylamine,2-chloro-5-nitro-4-pyrimidinamine,4-pyrimidinamine,2-chloro-5-nitro,acmc-209x0o,4-amino-5-nitro-2-chloropyrimidine,2-chloro-5-nitropyrimidine-4-amine,4-amino-2-chloro-5-nitropyrimidine
74716
2-chloro-5-nitropyrimidin-4-amine
C1=C(C(=NC(=N1)Cl)N)[N+](=O)[O-]
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Melting Point 221°C to 226°C
Beilstein 156953
Formula Weight 174.54
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-Amino-2-chloro-5-nitropyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning