Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-chloropyridine, 98%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
25g
14432-12-3
C5H5ClN2
128.559
MFCD00060089
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
14432-12-3
C5H5ClN2
128.559
MFCD00060089
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
Melting Point 91°C to 94°C
Boiling Point 153°C (5mmHg)
Quantity 1g
Packaging Plastic bottle
Beilstein 108671
Solubility Information Slightly soluble in water.
Formula Weight 128.56
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Amino-2-chloropyridine

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 238-403-0

TSCA : No

Recommended Storage : Ambient temperatures