Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-hydroxypyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
38767-72-5
C5H6N2O
110.12
SBQVQYPJWLJRQT-UHFFFAOYSA-N
4-aminopyridin-2-ol,4-amino-2-hydroxypyridine,4-amino-pyridin-2-ol,4-aminopyridin-2 1h-one,4-amino-2-pyridone,3-deazacytosine,2 1h-pyridinone, 4-amino,2 1h-pyridinone,4-amino,2-pyridinol, 4-amino,4-amino-1,2-dihydropyridin-2-one
573530
4-amino-1H-pyridin-2-one
C1=CNC(=O)C=C1N
38767-72-5
C5H6N2O
110.12
SBQVQYPJWLJRQT-UHFFFAOYSA-N
4-aminopyridin-2-ol,4-amino-2-hydroxypyridine,4-amino-pyridin-2-ol,4-aminopyridin-2 1h-one,4-amino-2-pyridone,3-deazacytosine,2 1h-pyridinone, 4-amino,2 1h-pyridinone,4-amino,2-pyridinol, 4-amino,4-amino-1,2-dihydropyridin-2-one
573530
4-amino-1H-pyridin-2-one
C1=CNC(=O)C=C1N
Melting Point 206.5°C to 208.5°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 96% min. (HPLC)
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Amino-2-hydroxypyridine

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou

GHS Signal Word: Danger