Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%

Click to view available options
Quantity:
1g
5g
25g
770-30-9
C6H6N4S
166.202
MFCD00023237
VYDHBDPZNLVELI-UHFFFAOYSA-N
4-amino-2-methylthio pyrimidine-5-carbonitrile,4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile,4-amino-2-methylthio-5-pyrimidinecarbonitrile,4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile,4-amino-2-methylthiopyrimidine-5-carbonitrile,maybridge1_004789,acmc-20a4j5,4-amino-5-cyano-2-methylthiopyrimidine,4-amino-5-cyano-2-methylthio pyrimidine
295757
4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
CSC1=NC=C(C(=N1)N)C#N
770-30-9
C6H6N4S
166.202
MFCD00023237
VYDHBDPZNLVELI-UHFFFAOYSA-N
4-amino-2-methylthio pyrimidine-5-carbonitrile,4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile,4-amino-2-methylthio-5-pyrimidinecarbonitrile,4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile,4-amino-2-methylthiopyrimidine-5-carbonitrile,maybridge1_004789,acmc-20a4j5,4-amino-5-cyano-2-methylthiopyrimidine,4-amino-5-cyano-2-methylthio pyrimidine
295757
4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
CSC1=NC=C(C(=N1)N)C#N
Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 5g
Melting Point 240°C to 243°C
UN Number UN3439
Solubility Information Soluble in water.
Formula Weight 166.21
Percent Purity 97%
Chemical Name or Material 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures