Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
770-30-9
C6H6N4S
166.21
MFCD00023237
VYDHBDPZNLVELI-UHFFFAOYSA-N
4-amino-2-methylthio pyrimidine-5-carbonitrile,4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile,4-amino-2-methylthio-5-pyrimidinecarbonitrile,4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile,4-amino-2-methylthiopyrimidine-5-carbonitrile,maybridge1_004789,acmc-20a4j5,4-amino-5-cyano-2-methylthiopyrimidine,4-amino-5-cyano-2-methylthio pyrimidine
295757
4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
CSC1=NC=C(C(=N1)N)C#N
770-30-9
C6H6N4S
166.21
MFCD00023237
VYDHBDPZNLVELI-UHFFFAOYSA-N
4-amino-2-methylthio pyrimidine-5-carbonitrile,4-amino-2-methylsulfanyl pyrimidine-5-carbonitrile,4-amino-2-methylthio-5-pyrimidinecarbonitrile,4-amino-2-methylsulfanyl-pyrimidine-5-carbonitrile,4-amino-2-methylthiopyrimidine-5-carbonitrile,maybridge1_004789,acmc-20a4j5,4-amino-5-cyano-2-methylthiopyrimidine,4-amino-5-cyano-2-methylthio pyrimidine
295757
4-amino-2-methylsulfanylpyrimidine-5-carbonitrile
CSC1=NC=C(C(=N1)N)C#N
Melting Point 240°C to 242°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Cream to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 25, V,17, 157
Formula Weight 166.21
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning