Alfa Aesar™ 4-Amino-2-nitrophenol, 97%

Manufacturer: Alfa Aesar™ H2769414

Quantity 25g

 View more versions of this product

Catalog No. AAH2769414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 119-34-6
    Molecular Formula C6H6N2O3
    Formula Weight 154.13
    MDL Number MFCD00007876
    Beilstein 1368435
    Synonym 4-Hydroxy-3-nitroaniline; C.I. 76555
    Chemical Name or Material 4-Amino-2-nitrophenol

    Specifications

    Formula Weight 154.13
    TSCA TSCA
    Melting Point 127° to 130°C
    Quantity 25g
    Molecular Formula C6H6N2O3
    MDL Number MFCD00007876
    CAS 119-34-6
    Beilstein 1368435
    Synonym 4-Hydroxy-3-nitroaniline; C.I. 76555
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H341-H302-H312-H335-H315
    Causes serious eye damage.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
    RTECS Number SJ6303000
    Chemical Name or Material 4-Amino-2-nitrophenol
    Assay 97%

    Description

    Applications
    4-Amino-2-nitrophenol was used in determination of aminonitrophenols in hair dyes by differential pulse voltammetry and HPLC with electrochemical detection. It is commonly used as semipermanent (nonoxidative) hair colorant and toner in permanent (oxidative) hair dye products.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool dry conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H318-H341-H302-H312-H335-H315
    Causes serious eye damage.
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P280-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit