Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3,5-dichlorobenzonitrile, 99%, Acros Organics

Click to view available options
Quantity:
1g
78473-00-4
C7H4Cl2N2
187.023
MFCD00017341
COFNCCWGWXFACE-UHFFFAOYSA-N
2,6-dichloro-4-cyanoaniline,4-cyano-2,6-dichloroaniline,benzonitrile, 4-amino-3,5-dichloro,3,5-dichloro-4-aminobenzonitrile,4-amino-3,5-dichloro-benzonitrile,4-amino-3,5-dichlorobenzenecarbonitrile,pubchem14453,acmc-1be7s,ksc381a4h,timtec-bb sbb003696
522706
4-amino-3,5-dichlorobenzonitrile
C1=C(C=C(C(=C1Cl)N)Cl)C#N
78473-00-4
C7H4Cl2N2
187.023
MFCD00017341
COFNCCWGWXFACE-UHFFFAOYSA-N
2,6-dichloro-4-cyanoaniline,4-cyano-2,6-dichloroaniline,benzonitrile, 4-amino-3,5-dichloro,3,5-dichloro-4-aminobenzonitrile,4-amino-3,5-dichloro-benzonitrile,4-amino-3,5-dichlorobenzenecarbonitrile,pubchem14453,acmc-1be7s,ksc381a4h,timtec-bb sbb003696
522706
4-amino-3,5-dichlorobenzonitrile
C1=C(C=C(C(=C1Cl)N)Cl)C#N
Quantity 1g
Melting Point 116°C to 120°C
Infrared Spectrum Authentic
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 187.02
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Amino-3, 5-dichlorobenzonitrile, 99%
Show More Show Less