Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3,5-dichloropyridine, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
100g
22889-78-7
C5H4Cl2N2
163.01
ISIQAMHROGZHOV-UHFFFAOYSA-N
4-amino-3,5-dichloropyridine,3,5-dichloro-4-aminopyridine,3,5-dichloro-4-pyridinamine,4-pyridinamine, 3,5-dichloro,3,5-dichloro-4-amino pyridine,3,5-dichloro-4-pyridine amine,3,5-dichloro-pyridin-4-ylamine,3,5-dichloro-4-pyridylamine,3,5-dichloropyridin-4-ylamine,4-amino-3,5-dichloro pyridine
89888
3,5-dichloropyridin-4-amine
C1=C(C(=C(C=N1)Cl)N)Cl
22889-78-7
C5H4Cl2N2
163.01
ISIQAMHROGZHOV-UHFFFAOYSA-N
4-amino-3,5-dichloropyridine,3,5-dichloro-4-aminopyridine,3,5-dichloro-4-pyridinamine,4-pyridinamine, 3,5-dichloro,3,5-dichloro-4-amino pyridine,3,5-dichloro-4-pyridine amine,3,5-dichloro-pyridin-4-ylamine,3,5-dichloro-4-pyridylamine,3,5-dichloropyridin-4-ylamine,4-amino-3,5-dichloro pyridine
89888
3,5-dichloropyridin-4-amine
C1=C(C(=C(C=N1)Cl)N)Cl
Quantity 25g
Melting Point 158.0°C to 160.0°C
Infrared Spectrum Authentic
Water 1% max.
Assay Percent Range 96% min. (GC)
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 4-Amino-3,5-dichloropyridine

GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 245-304-6