Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3,5-dichloropyridine, 98+%

Click to view available options
Quantity:
5g
25g
100g
22889-78-7
C5H4Cl2N2
163.001
MFCD00125023
ISIQAMHROGZHOV-UHFFFAOYSA-N
4-amino-3,5-dichloropyridine,3,5-dichloro-4-aminopyridine,3,5-dichloro-4-pyridinamine,4-pyridinamine, 3,5-dichloro,3,5-dichloro-4-amino pyridine,3,5-dichloro-4-pyridine amine,3,5-dichloro-pyridin-4-ylamine,3,5-dichloro-4-pyridylamine,3,5-dichloropyridin-4-ylamine,4-amino-3,5-dichloro pyridine
89888
3,5-dichloropyridin-4-amine
C1=C(C(=C(C=N1)Cl)N)Cl
22889-78-7
C5H4Cl2N2
163.001
MFCD00125023
ISIQAMHROGZHOV-UHFFFAOYSA-N
4-amino-3,5-dichloropyridine,3,5-dichloro-4-aminopyridine,3,5-dichloro-4-pyridinamine,4-pyridinamine, 3,5-dichloro,3,5-dichloro-4-amino pyridine,3,5-dichloro-4-pyridine amine,3,5-dichloro-pyridin-4-ylamine,3,5-dichloro-4-pyridylamine,3,5-dichloropyridin-4-ylamine,4-amino-3,5-dichloro pyridine
89888
3,5-dichloropyridin-4-amine
C1=C(C(=C(C=N1)Cl)N)Cl
Melting Point 159°C to 163°C
Quantity 25g
Beilstein 120547
Formula Weight 163.01
Percent Purity ≥98%
Chemical Name or Material 4-Amino-3,5-dichloropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning