Learn More
4-Amino-3,5-diiodobenzoic acid, tech. 90%, Thermo Scientific Chemicals
Description
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.
Solubility
Soluble in water (partly miscible).
Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 2103037 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in water (partly miscible). |
Formula Weight | 388.93 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 4-Amino-3,5-diiodobenzoic acid |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 218-331-6 |
RTECSNumber | DG2310000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.