Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3,5-diiodobenzoic acid, tech. 90%

Manufacturer:  Alfa Aesar™ A1061706

Quantity 5g

 View more versions of this product

Catalog No. AAA1061706


  Check Availability
Add to cart

Specifications

Description

Applications
4-Amino-3,5-diiodobenzoic acid is a very useful substituted benzene compound for use as a building block in synthesis. The carboxylic acid and amine functionalities can be used to form linkages through nucleophile-electrophile coupling and the iodides may be used as functional sites for cross-coupling transformations. 4-Amino-3,5-diiodobenzoic acid has been specifically used in the synthesis of an adduct to introduce into Tecoflex polymers to impart radiopaqueness to the material.

Solubility
Soluble in water (partly miscible).

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Description & Specifications

Chemical Identifiers

2122-61-4
388.93
2103037
Technical Grade
C7H5I2NO2
MFCD00007684
4-Amino-3,5-diiodobenzoic acid

Specifications

2122-61-4
5g
388.93
2103037
90%
>300°C
C7H5I2NO2
MFCD00007684
4-Amino-3,5-diiodobenzoic acid
Technical Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG2310000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit