Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-bromobenzonitrile 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
50397-74-5
C7H5BrN2
197.035
MFCD01861386
POESQIHWIIWNJL-UHFFFAOYSA-N
3-bromo-4-aminobenzonitrile,2-bromo-4-cyanoaniline,benzonitrile, 4-amino-3-bromo,4-amino-3-bromo-benzonitrile,4-amino-3-brombenzonitrile,buttpark 35\03-64,4-amino-3-bromobenzenecarbonitrile,pubchem4609,acmc-209klz,4-amino-3-bromobenzonitile
1515279
4-amino-3-bromobenzonitrile
C1=CC(=C(C=C1C#N)Br)N
50397-74-5
C7H5BrN2
197.035
MFCD01861386
POESQIHWIIWNJL-UHFFFAOYSA-N
3-bromo-4-aminobenzonitrile,2-bromo-4-cyanoaniline,benzonitrile, 4-amino-3-bromo,4-amino-3-bromo-benzonitrile,4-amino-3-brombenzonitrile,buttpark 35\03-64,4-amino-3-bromobenzenecarbonitrile,pubchem4609,acmc-209klz,4-amino-3-bromobenzonitile
1515279
4-amino-3-bromobenzonitrile
C1=CC(=C(C=C1C#N)Br)N
Color Yellow
Melting Point 112°C
Quantity 25g
UN Number 3439
Formula Weight 197.04
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,N)
Chemical Name or Material 4-Amino-3-bromobenzonitrile