Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-bromobenzotrifluoride, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
57946-63-1
C7H5BrF3N
240.02
MFCD00042150
QKRJIXSZTKOFTD-UHFFFAOYSA-N
2-bromo-4-trifluoromethyl aniline,4-amino-3-bromobenzotrifluoride,2-bromo-4-trifluoromethyl-phenylamine,benzenamine, 2-bromo-4-trifluoromethyl,3-bromo-4-aminobenzotrifluoride,4-amino-3-bromo benzotrifluoride,buttpark 83\07-26,timtec-bb sbb000789,buttpark 44\03-51,2-bromo-4-trifluoromethyl phenylamine
93899
2-bromo-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)Br)N
57946-63-1
C7H5BrF3N
240.02
MFCD00042150
QKRJIXSZTKOFTD-UHFFFAOYSA-N
2-bromo-4-trifluoromethyl aniline,4-amino-3-bromobenzotrifluoride,2-bromo-4-trifluoromethyl-phenylamine,benzenamine, 2-bromo-4-trifluoromethyl,3-bromo-4-aminobenzotrifluoride,4-amino-3-bromo benzotrifluoride,buttpark 83\07-26,timtec-bb sbb000789,buttpark 44\03-51,2-bromo-4-trifluoromethyl phenylamine
93899
2-bromo-4-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)Br)N
Melting Point 32°C to 35°C
Color White
Boiling Point 110°C (10.0mmHg)
Flash Point 110°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 240.02
Physical Form Crystals
Percent Purity 99%
Chemical Name or Material 4-Amino-3-bromobenzotrifluoride
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 261-035-7