Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-chlorobenzonitrile 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
21803-75-8
C7H5ClN2
152.581
MFCD00052917
OREVCMGFYSUYPX-UHFFFAOYSA-N
3-chloro-4-aminobenzonitrile,2-chloro-4-cyanoaniline,3-chloro-4-aminbenzonitrile,4-amino-3-chloro-benzonitrile,benzonitrile, 4-amino-3-chloro,4-amino-3-chlorobenzenecarbonitrile,pubchem4625,2-chloro-4-cyanoanilin,2-chloro-4-cyano-aniline,acmc-1cs04
519896
4-amino-3-chlorobenzonitrile
C1=CC(=C(C=C1C#N)Cl)N
21803-75-8
C7H5ClN2
152.581
MFCD00052917
OREVCMGFYSUYPX-UHFFFAOYSA-N
3-chloro-4-aminobenzonitrile,2-chloro-4-cyanoaniline,3-chloro-4-aminbenzonitrile,4-amino-3-chloro-benzonitrile,benzonitrile, 4-amino-3-chloro,4-amino-3-chlorobenzenecarbonitrile,pubchem4625,2-chloro-4-cyanoanilin,2-chloro-4-cyano-aniline,acmc-1cs04
519896
4-amino-3-chlorobenzonitrile
C1=CC(=C(C=C1C#N)Cl)N
Color Undesignated
Quantity 25g
Melting Point 104°C
UN Number 3439
Formula Weight 152.58
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 4-Amino-3-chlorobenzonitrile