Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3-chlorophenol, 95%

Click to view available options
Quantity:
250mg
1g
17609-80-2
C6H6ClNO
143.57
MFCD00801116
PNLPXABQLXSICH-UHFFFAOYSA-N
3-chloro-4-aminophenol,phenol, 4-amino-3-chloro,2-chloro-4-hydroxyaniline,4-amino-3-chloro-phenol,amino 4--3-chlorophenol,pubchem4513,4-amino-3-chloro-pheno,3-13-00-01184 beilstein handbook reference,3-chloro-4-aminophenol;4-amino-3-chloro-pheno,l-valine ethyl ester hydrochloride; ethyl 2-amino-3-methylbutanoate hydrochloride
87184
4-amino-3-chlorophenol
C1=CC(=C(C=C1O)Cl)N
17609-80-2
C6H6ClNO
143.57
MFCD00801116
PNLPXABQLXSICH-UHFFFAOYSA-N
3-chloro-4-aminophenol,phenol, 4-amino-3-chloro,2-chloro-4-hydroxyaniline,4-amino-3-chloro-phenol,amino 4--3-chlorophenol,pubchem4513,4-amino-3-chloro-pheno,3-13-00-01184 beilstein handbook reference,3-chloro-4-aminophenol;4-amino-3-chloro-pheno,l-valine ethyl ester hydrochloride; ethyl 2-amino-3-methylbutanoate hydrochloride
87184
4-amino-3-chlorophenol
C1=CC(=C(C=C1O)Cl)N
Color White
Quantity 250mg
Melting Point 159°C to 160°C
UN Number UN2811
Beilstein 2802558
Formula Weight 143.57
Physical Form Crystalline Powder
Percent Purity 95%
Chemical Name or Material 4-Amino-3-chlorophenol

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : SJ5710000