Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-fluorophenylboronic acid hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
1256355-32-4
C6H7BFNO2·ClH
191.4
NXCNVCKJELIVDM-UHFFFAOYSA-N
4-amino-3-fluorophenylboronic acid hydrochloride,4-amino-3-fluorophenylboronic acid, hydrochloride,4-amino-3-fluorophenyl boronic acid hydrochloride,4-amino-3-fluorophenylboronic acid,hydrochloride,4-amino-3-fluorophenylboronic acid,hcl,4-amino-3-fluorophenylboronic acid, hcl,4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1,boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
46738020
1256355-32-4
C6H7BFNO2·ClH
191.4
NXCNVCKJELIVDM-UHFFFAOYSA-N
4-amino-3-fluorophenylboronic acid hydrochloride,4-amino-3-fluorophenylboronic acid, hydrochloride,4-amino-3-fluorophenyl boronic acid hydrochloride,4-amino-3-fluorophenylboronic acid,hydrochloride,4-amino-3-fluorophenylboronic acid,hcl,4-amino-3-fluorophenylboronic acid, hcl,4-amino-3-fluorophenyl boronic acid-hydrogen chloride 1/1,boronic acid, b-4-amino-3-fluorophenyl-, hydrochloride 1:1
46738020
Chloride 18 to 19%
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 191.4
Percent Purity 97%
Chemical Name or Material 4-Amino-3-fluorophenylboronic acid hydrochloride

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning