Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxy-1-naphthalenesulfonic Acid 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
116-63-2
C10H9NO4S
239.25
MFCD00004019
RXCMFQDTWCCLBL-UHFFFAOYSA-N
1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine
8316
CHEBI:19024
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
116-63-2
C10H9NO4S
239.25
MFCD00004019
RXCMFQDTWCCLBL-UHFFFAOYSA-N
1-amino-2-naphthol-4-sulfonic acid,4-amino-3-hydroxy-1-naphthalenesulfonic acid,1-amino-4-sulfo-2-naphthol,1,2,4-acid,unii-w46sk3p33g,1-naphthalenesulfonic acid, 4-amino-3-hydroxy,4-amino-3-hydroxynaphthalene-1-sulphonic acid,4-amino-3-hydroxynaphthalenesulfonic acid,1,2,4-aminonaphtholsulfonic acid,2-hydroxy-4-sulfo-1-naphthylamine
8316
CHEBI:19024
4-amino-3-hydroxynaphthalene-1-sulfonic acid
C1=CC=C2C(=C1)C(=CC(=C2N)O)S(=O)(=O)O
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
Sulfated Ash 0.5% max.
Color Pink or Purple
Melting Point 295.0°C
Water 5% max. (K.F.)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula H2NC10H5(OH)SO3H
Beilstein 14, 846
Merck Index 15, 450
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot sodium bisulfite sol., soluble in alkaline solutions, insoluble in alcohol, ether and benzene
Formula Weight 239.25
Physical Form Granular Powder
Percent Purity 95%
Chemical Name or Material 4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 204-147-3

RTECSNumber : QK1292000

TSCA : TSCA