Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-hydroxybenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2374-03-0
C7H7NO3
153.14
MFCD00017094
NFPYJDZQOKCYIE-UHFFFAOYSA-N
3-hydroxy-4-aminobenzoic acid,4-amino-3-hydroxy benzoic acid,benzoic acid, 4-amino-3-hydroxy,chembl1462,2hdr,pubchem14418,2-amino-5-carboxyphenol,intermediates-zcf02635,acmc-209g5k,4-carboxy-2-hydroxyaniline
137566
4-amino-3-hydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)N
2374-03-0
C7H7NO3
153.14
MFCD00017094
NFPYJDZQOKCYIE-UHFFFAOYSA-N
3-hydroxy-4-aminobenzoic acid,4-amino-3-hydroxy benzoic acid,benzoic acid, 4-amino-3-hydroxy,chembl1462,2hdr,pubchem14418,2-amino-5-carboxyphenol,intermediates-zcf02635,acmc-209g5k,4-carboxy-2-hydroxyaniline
137566
4-amino-3-hydroxybenzoic acid
C1=CC(=C(C=C1C(=O)O)O)N
Melting Point 211°C to 215°C
Color Brown to Brown-Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 14, IV, 2075
Solubility Information Solubility in water: .. Other solubilities: soluble in chcl3 and aq. ethanol
Formula Weight 153.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Amino-3-hydroxybenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning