Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3-methylbenzenesulfonic acid, 98%

Click to view available options
Quantity:
250g
1000g
98-33-9
C7H9NO3S
187.213
MFCD00025198
WQTCZINVPXJNEL-UHFFFAOYSA-N
2-aminotoluene-5-sulfonic acid,benzenesulfonic acid, 4-amino-3-methyl,4-amino-m-toluenesulfonic acid,2-amino-5-toluenesulfonic acid,o-toluidine-m-sulfonic acid,6-aminotoluene-3-sulfonic acid,3-methylsulphanilic acid,unii-et833cb2ku,ccris 3412,4-amino-m-toluenesulfonic acid so3h=1
7384
4-amino-3-methylbenzenesulfonic acid
CC1=C(C=CC(=C1)S(=O)(=O)O)N
98-33-9
C7H9NO3S
187.213
MFCD00025198
WQTCZINVPXJNEL-UHFFFAOYSA-N
2-aminotoluene-5-sulfonic acid,benzenesulfonic acid, 4-amino-3-methyl,4-amino-m-toluenesulfonic acid,2-amino-5-toluenesulfonic acid,o-toluidine-m-sulfonic acid,6-aminotoluene-3-sulfonic acid,3-methylsulphanilic acid,unii-et833cb2ku,ccris 3412,4-amino-m-toluenesulfonic acid so3h=1
7384
4-amino-3-methylbenzenesulfonic acid
CC1=C(C=CC(=C1)S(=O)(=O)O)N
Odor Odorless
Quantity 250g
Melting Point >300°C
Formula Weight 187.22
Percent Purity 98%
Chemical Name or Material 4-Amino-3-methylbenzenesulfonic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c

H317-H319

EINECSNumber : 202-658-6

TSCA : Yes

Recommended Storage : Ambient temperatures