Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 4-Amino-3-methylbenzoic acid, 98%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Melting Point | 169°C to 171°C |
Quantity | 25g |
Beilstein | 2802615 |
Formula Weight | 151.16 |
Percent Purity | 98% |
Chemical Name or Material | 4-Amino-3-methylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-629-9
TSCA : No
Recommended Storage : Ambient temperatures