Alfa Aesar™ 4-Amino-3-methylphenol, 98%

Manufacturer: Alfa Aesar™ B2315214

Quantity 25g

 View more versions of this product

Catalog No. AAB2315214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2835-99-6
    Molecular Formula C7H9NO
    Formula Weight 123.16
    MDL Number MFCD00007871
    UN Number UN3077
    Beilstein 2078803
    Synonym 4-Amino-m-cresol; 4-Hydroxy-2-methylaniline
    Chemical Name or Material 4-Amino-3-methylphenol

    Specifications

    Formula Weight 123.16
    Melting Point 176° to 179°C
    Quantity 25g
    MDL Number MFCD00007871
    UN Number UN3077
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Molecular Formula C7H9NO
    CAS 2835-99-6
    Beilstein 2078803
    Synonym 4-Amino-m-cresol; 4-Hydroxy-2-methylaniline
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number GO6826000
    Chemical Name or Material 4-Amino-3-methylphenol
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit