Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-3-nitrobenzonitrile, 98%

Click to view available options
Quantity:
1g
5g
25g
6393-40-4
C7H5N3O2
163.136
MFCD00013373
JAHADAZIDZMHOP-UHFFFAOYSA-N
3-nitro-4-aminobenzonitrile,4-cyano-2-nitroaniline,benzonitrile, 4-amino-3-nitro,4-amino-3-nitro-benzonitrile,2-nitro-4-cyanoaniline,4-amino-3-nitrobenzenecarbonitrile,zlchem 228,pubchem4669,4-ammo-3-nitrobenzonitrile
595901
4-amino-3-nitrobenzonitrile
C1=CC(=C(C=C1C#N)[N+](=O)[O-])N
6393-40-4
C7H5N3O2
163.136
MFCD00013373
JAHADAZIDZMHOP-UHFFFAOYSA-N
3-nitro-4-aminobenzonitrile,4-cyano-2-nitroaniline,benzonitrile, 4-amino-3-nitro,4-amino-3-nitro-benzonitrile,2-nitro-4-cyanoaniline,4-amino-3-nitrobenzenecarbonitrile,zlchem 228,pubchem4669,4-ammo-3-nitrobenzonitrile
595901
4-amino-3-nitrobenzonitrile
C1=CC(=C(C=C1C#N)[N+](=O)[O-])N
Applications
4-Amino-3-nitrobenzonitrile is used in gamma irradiations effect on release of this compound from polyanhydride matrixes.

Solubility
Soluble in ethanol and benzene.

Notes
Incompatible with strong oxidizing agents.
Melting Point 159°C to 161°C
Quantity 1g
UN Number UN3439
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitrobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.