Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-nitrobenzonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6393-40-4
C7H5N3O2
163.136
MFCD00013373
JAHADAZIDZMHOP-UHFFFAOYSA-N
3-nitro-4-aminobenzonitrile,4-cyano-2-nitroaniline,benzonitrile, 4-amino-3-nitro,4-amino-3-nitro-benzonitrile,2-nitro-4-cyanoaniline,4-amino-3-nitrobenzenecarbonitrile,zlchem 228,pubchem4669,4-ammo-3-nitrobenzonitrile
595901
4-amino-3-nitrobenzonitrile
C1=CC(=C(C=C1C#N)[N+](=O)[O-])N
6393-40-4
C7H5N3O2
163.136
MFCD00013373
JAHADAZIDZMHOP-UHFFFAOYSA-N
3-nitro-4-aminobenzonitrile,4-cyano-2-nitroaniline,benzonitrile, 4-amino-3-nitro,4-amino-3-nitro-benzonitrile,2-nitro-4-cyanoaniline,4-amino-3-nitrobenzenecarbonitrile,zlchem 228,pubchem4669,4-ammo-3-nitrobenzonitrile
595901
4-amino-3-nitrobenzonitrile
C1=CC(=C(C=C1C#N)[N+](=O)[O-])N
Packaging Glass bottle
Melting Point 159°C to 163°C
Color Yellow
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula NCC6H3(NO2)NH2
Formula Weight 163.14
Physical Form Powder, Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Amino-3-nitrobenzonitrile
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning