Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-3-nitropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1681-37-4
C5H5N3O2
139.114
MFCD00160304
IUPPEELMBOPLDJ-UHFFFAOYSA-N
4-amino-3-nitropyridine,4-pyridinamine, 3-nitro,3-nitro-4-pyridinamine,3-nitro-4-aminopyridine,3-nitro-pyridin-4-ylamine,3-nitro-4-pyridylamine,pubchem1277,acmc-209dxo,3-nitro-4-amino pyridine,3-nitro-4-pyridinamine #
548803
3-nitropyridin-4-amine
C1=CN=CC(=C1N)[N+](=O)[O-]
1681-37-4
C5H5N3O2
139.114
MFCD00160304
IUPPEELMBOPLDJ-UHFFFAOYSA-N
4-amino-3-nitropyridine,4-pyridinamine, 3-nitro,3-nitro-4-pyridinamine,3-nitro-4-aminopyridine,3-nitro-pyridin-4-ylamine,3-nitro-4-pyridylamine,pubchem1277,acmc-209dxo,3-nitro-4-amino pyridine,3-nitro-4-pyridinamine #
548803
3-nitropyridin-4-amine
C1=CN=CC(=C1N)[N+](=O)[O-]
Quantity 1g
Melting Point 195.0°C to 200.0°C
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 22, V,9, 188
Formula Weight 139.11
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Amino-3-nitropyridine
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning