Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Amino-4'-chlorodiphenyl ether, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is used as an active pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 100°C to 103°C |
Odor | Odorless |
Quantity | 10 g |
Solubility Information | Insoluble in water. |
Formula Weight | 219.67 |
Percent Purity | 97% |
Chemical Name or Material | 4-Amino-4'-chlorodiphenyl ether |
Safety and Handling
Hazard Category | H315-H319-H335-H351 |
Hazard Statement | GHS H Statement H351-H315-H319-H335 Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
EINECSNumber | 202-976-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction