Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-5,6-dichloropyrimidine, 98%

Click to view available options
Quantity:
1g
5g
25g
310400-38-5
C4H3Cl2N3
163.989
MFCD09837293
ISTJUVRUUBNZNB-UHFFFAOYSA-N
4-amino-5,6-dichloropyrimidine,5,6-dichloro-pyrimidin-4-ylamine,4-pyrimidinamine, 5,6-dichloro,5,6-dichloro-4-pyrimidinamine,5,6-dichloropyrimidin-4-ylamine,4-pyrimidinamine, 5,6-dichloro-9ci
24219262
5,6-dichloropyrimidin-4-amine
C1=NC(=C(C(=N1)Cl)Cl)N
310400-38-5
C4H3Cl2N3
163.989
MFCD09837293
ISTJUVRUUBNZNB-UHFFFAOYSA-N
4-amino-5,6-dichloropyrimidine,5,6-dichloro-pyrimidin-4-ylamine,4-pyrimidinamine, 5,6-dichloro,5,6-dichloro-4-pyrimidinamine,5,6-dichloropyrimidin-4-ylamine,4-pyrimidinamine, 5,6-dichloro-9ci
24219262
5,6-dichloropyrimidin-4-amine
C1=NC(=C(C(=N1)Cl)Cl)N
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Color White
Quantity 1g
Beilstein 8678505
Formula Weight 163.99
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Amino-5,6-dichloropyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning