Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-5-bromo-6-chloropyrimidine, 98%

Click to view available options
Quantity:
250mg
1g
5g
663193-80-4
C4H3BrClN3
208.443
MFCD09999224
DQSBRYJETNYGCI-UHFFFAOYSA-N
4-amino-5-bromo-6-chloropyrimidine,5-bromo-6-chloro-4-pyrimidinamine,5-bromo-6-chloro-pyrimidin-4-ylamine
45789713
5-bromo-6-chloropyrimidin-4-amine
C1=NC(=C(C(=N1)Cl)Br)N
663193-80-4
C4H3BrClN3
208.443
MFCD09999224
DQSBRYJETNYGCI-UHFFFAOYSA-N
4-amino-5-bromo-6-chloropyrimidine,5-bromo-6-chloro-4-pyrimidinamine,5-bromo-6-chloro-pyrimidin-4-ylamine
45789713
5-bromo-6-chloropyrimidin-4-amine
C1=NC(=C(C(=N1)Cl)Br)N
Color Gray
Quantity 1g
Beilstein 23483365
Formula Weight 208.44
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 4-Amino-5-bromo-6-chloropyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning