Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-5-bromopyrimidine, 98%

Click to view available options
Quantity:
250mg
1g
5g
1439-10-7
C4H4BrN3
174.001
MFCD00233962
IIFAONYUCDAVGA-UHFFFAOYSA-N
4-amino-5-bromopyrimidine,5-bromo-pyrimidin-4-ylamine,4-pyrimidinamine, 5-bromo,4-pyrimidinamine 5-bromo,5-bromopyrimidine-4-ylamine,pubchem14978,acmc-209crn,4-pyrimidinamine,5-bromo,4-amino-5-bromo-pyrimidine,pyrimidine, 4-amino-5-bromo
10261576
5-bromopyrimidin-4-amine
C1=C(C(=NC=N1)N)Br
1439-10-7
C4H4BrN3
174.001
MFCD00233962
IIFAONYUCDAVGA-UHFFFAOYSA-N
4-amino-5-bromopyrimidine,5-bromo-pyrimidin-4-ylamine,4-pyrimidinamine, 5-bromo,4-pyrimidinamine 5-bromo,5-bromopyrimidine-4-ylamine,pubchem14978,acmc-209crn,4-pyrimidinamine,5-bromo,4-amino-5-bromo-pyrimidine,pyrimidine, 4-amino-5-bromo
10261576
5-bromopyrimidin-4-amine
C1=C(C(=NC=N1)N)Br
Applications
It is an inhibitor of thymidine phosphorylase, an enzyme known to be over-expressed in several tumor types and to play a role in metastasis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
Color Brown
Melting Point 207°C to 211°C
Beilstein 112648
Formula Weight 174
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4-Amino-5-bromopyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning