Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-5-chloro-2,6-dimethylpyrimidine, 99%

Click to view available options
Quantity:
5g
25g
2858-20-0
C6H8ClN3
157.601
MFCD00191727
BKFCZKYCVQQMCX-UHFFFAOYSA-N
4-amino-5-chloro-2,6-dimethylpyrimidine,timtec-bb sbb004000,4-amino-2-chloro-6,7-dimethylpyrimidine,5-chloro-2,6-dimethylpyrimidine-4-ylamine,acmc-209h3e,5-chloro-2,6-dimethyl-4-pyrimidinamine,2,4-dimethyl-5-chloro-6-aminopyrimidine,2,6-dimethyl-4-amino-5-chloropyrimidine,4-pyrimidinamine,5-chloro-2,6-dimethyl,5-chloro-2,6-dimethyl-4-pyrimidinylamine
593843
5-chloro-2,6-dimethylpyrimidin-4-amine
CC1=C(C(=NC(=N1)C)N)Cl
2858-20-0
C6H8ClN3
157.601
MFCD00191727
BKFCZKYCVQQMCX-UHFFFAOYSA-N
4-amino-5-chloro-2,6-dimethylpyrimidine,timtec-bb sbb004000,4-amino-2-chloro-6,7-dimethylpyrimidine,5-chloro-2,6-dimethylpyrimidine-4-ylamine,acmc-209h3e,5-chloro-2,6-dimethyl-4-pyrimidinamine,2,4-dimethyl-5-chloro-6-aminopyrimidine,2,6-dimethyl-4-amino-5-chloropyrimidine,4-pyrimidinamine,5-chloro-2,6-dimethyl,5-chloro-2,6-dimethyl-4-pyrimidinylamine
593843
5-chloro-2,6-dimethylpyrimidin-4-amine
CC1=C(C(=NC(=N1)C)N)Cl
Quantity 5g
Melting Point 164°C to 166°C
Formula Weight 157.6
Percent Purity 99%
Chemical Name or Material 4-Amino-5-chloro-2,6-dimethylpyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning