Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Amino-5-chloro-2-methoxybenzoic acid, 98+%

Click to view available options
Quantity:
5g
25g
7206-70-4
C8H8ClNO3
201.606
MFCD00038794
RVEATKYEARPWRE-UHFFFAOYSA-N
4-amino-5-chloro-o-anisic acid,unii-pq8l2l84el,benzoic acid, 4-amino-5-chloro-2-methoxy,pq8l2l84el,pubchem15052,acmc-209oll,metoclopramide impurity c,2-methoxy-4-amino-5-chlorobenzoic acid,4amino-5-chloro-2-methoxy-benzoic acid,5-chloro-4-amino-2-methoxybenzoic acid
81626
4-amino-5-chloro-2-methoxybenzoic acid
COC1=CC(=C(C=C1C(=O)O)Cl)N
7206-70-4
C8H8ClNO3
201.606
MFCD00038794
RVEATKYEARPWRE-UHFFFAOYSA-N
4-amino-5-chloro-o-anisic acid,unii-pq8l2l84el,benzoic acid, 4-amino-5-chloro-2-methoxy,pq8l2l84el,pubchem15052,acmc-209oll,metoclopramide impurity c,2-methoxy-4-amino-5-chlorobenzoic acid,4amino-5-chloro-2-methoxy-benzoic acid,5-chloro-4-amino-2-methoxybenzoic acid
81626
4-amino-5-chloro-2-methoxybenzoic acid
COC1=CC(=C(C=C1C(=O)O)Cl)N
Quantity 25g
Melting Point 205°C to 208°C (decomposition)
Formula Weight 201.61
Percent Purity ≥98%
Chemical Name or Material 4-Amino-5-chloro-2-methoxybenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning