Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-imidazolecarboxamide hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
72-40-2,360-97-4
C4H6N4O·HCl
162.58
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride,5-amino-4-imidazolecarboxamide hydrochloride,5-amino-1h-imidazole-4-carboxamide hydrochloride,5-aminoimidazole-4-carboxamide hydrochloride,aic .cntdot. hcl,4-amino-1h-imidazole-5-carboxamide hydrochloride,aminoimidazolecarboxamide hydrochloride,4-aminoimidazole-5-carboxamide hydrochoride,1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride,5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
C1=NC(=C(N1)C(=O)N)N.Cl
72-40-2,360-97-4
C4H6N4O·HCl
162.58
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride,5-amino-4-imidazolecarboxamide hydrochloride,5-amino-1h-imidazole-4-carboxamide hydrochloride,5-aminoimidazole-4-carboxamide hydrochloride,aic .cntdot. hcl,4-amino-1h-imidazole-5-carboxamide hydrochloride,aminoimidazolecarboxamide hydrochloride,4-aminoimidazole-5-carboxamide hydrochoride,1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride,5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
C1=NC(=C(N1)C(=O)N)N.Cl
Melting Point 250°C
Loss on Drying 0.5% max. (105°C)
Color White
Appearance (1% soln.) clear, colorless
Infrared Spectrum Authentic
Quantity 1g
Ignition Residue 0.2% max.
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Beilstein 25, II, 221
Solubility Information (5% in water) Clear
Formula Weight 162.58
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Amino-5-imidazolecarboxamide hydrochloride, 98%
Show More Show Less

EINECSNumber : 200-778-3

RTECSNumber : NI3911000

TSCA : TSCA