Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-5-imidazolecarboxamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number AC173090050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
72-40-2, 360-97-4
C4H7ClN4O
162.58
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
Cl.NC(=O)C1=C(N)N=CN1
72-40-2, 360-97-4
C4H7ClN4O
162.58
MFCD00012704
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
66146
4-amino-1H-imidazole-5-carboxamide;hydrochloride
Cl.NC(=O)C1=C(N)N=CN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color White
Ignition Residue 0.2% max.
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 25, II, 221
Solubility Information Solubility in water: Soluble
Formula Weight 162.58
Percent Purity ≥97.5% (Argentometry)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Amino-5-imidazolecarboxamide hydrochloride, 98%
Show More Show Less
EINECSNumber 200-778-3
RTECSNumber NI3911000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit