Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-6-methoxypyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
696-45-7
C5H7N3O
125.13
MFCD00129983
VELRBZDRGTVGGT-UHFFFAOYSA-N
4-amino-6-methoxypyrimidine,6-methoxy-pyrimidin-4-ylamine,6-methoxy-4-pyrimidinamine,4-amino-6-methoxy pyrimidine,4-pyrimidinamine, 6-methoxy,6-methoxypyrimidine-4-ylamine,6-methoxypyrimidin-4-amin,pubchem2605,acmc-209oa2,6-amino-4-methoxypyrimidine
735731
6-methoxypyrimidin-4-amine
COC1=NC=NC(=C1)N
696-45-7
C5H7N3O
125.13
MFCD00129983
VELRBZDRGTVGGT-UHFFFAOYSA-N
4-amino-6-methoxypyrimidine,6-methoxy-pyrimidin-4-ylamine,6-methoxy-4-pyrimidinamine,4-amino-6-methoxy pyrimidine,4-pyrimidinamine, 6-methoxy,6-methoxypyrimidine-4-ylamine,6-methoxypyrimidin-4-amin,pubchem2605,acmc-209oa2,6-amino-4-methoxypyrimidine
735731
6-methoxypyrimidin-4-amine
COC1=NC=NC(=C1)N
Infrared Spectrum Authentic
Quantity 25g
Color White to Yellow
Melting Point 154.0°C to 159.0°C
Packaging Glass Bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 125.13
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Amino-6-methoxypyrimidine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning