Alfa Aesar™ 4-Amino-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H51970MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51970MD

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 943-80-6
    Molecular Formula C9H12N2O2
    Formula Weight 180.2
    MDL Number MFCD00069927
    Synonym (S)-2-Amino-3-(4-aminophenyl)propionic acid; H-Phe(4-NH2)-OH
    Chemical Name or Material 4-Amino-L-phenylalanine

    Specifications

    Formula Weight 180.2
    Molecular Formula C9H12N2O2
    MDL Number MFCD00069927
    CAS 943-80-6
    Synonym (S)-2-Amino-3-(4-aminophenyl)propionic acid; H-Phe(4-NH2)-OH
    Quantity 250mg
    Chemical Name or Material 4-Amino-L-phenylalanine
    Assay 95%

    Description

    Applications
    It finds its application in the purification of peptide synthetases involved in pristinamycin I biosynthesis.

    Solubility
    Soluble in water.

    Notes
    Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit