Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-m-cresol, 99+%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
2835-99-6
C7H9NO
123.15
MFCD00007871
QGNGOGOOPUYKMC-UHFFFAOYSA-N
4-amino-m-cresol,4-hydroxy-2-methylaniline,3-methyl-4-aminophenol,p-amino-m-cresol,phenol, 4-amino-3-methyl,p-hydroxy-o-toluidine,2-methyl-4-hydroxyaniline,2-amino-5-hydroxytoluene,m-cresol, 4-amino,4-amino-3-cresol
17819
CHEBI:55546
4-amino-3-methylphenol
CC1=C(C=CC(=C1)O)N
2835-99-6
C7H9NO
123.15
MFCD00007871
QGNGOGOOPUYKMC-UHFFFAOYSA-N
4-amino-m-cresol,4-hydroxy-2-methylaniline,3-methyl-4-aminophenol,p-amino-m-cresol,phenol, 4-amino-3-methyl,p-hydroxy-o-toluidine,2-methyl-4-hydroxyaniline,2-amino-5-hydroxytoluene,m-cresol, 4-amino,4-amino-3-cresol
17819
CHEBI:55546
4-amino-3-methylphenol
CC1=C(C=CC(=C1)O)N
Quantity 50g
Melting Point 176°C to 180°C
Color Brown
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula H2NC6H3(CH3)OH
Beilstein 13, 593
Solubility Information Solubility in water: insoluble
Formula Weight 123.15
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material 4-Amino-m-cresol, 99+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-621-2

RTECSNumber : GO6826000

TSCA : TSCA