Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-N-(2-hydroxyethyl)-N-ethylaniline Sulfate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
4327-84-8
C10H18N2O5S
278.323
MFCD00040597
KAJALVWKFPQZOO-UHFFFAOYSA-N
N-Ethyl-N-(2-hydroxyethyl)-1,4-phenylenediamine Sulfate
2756529
2-(4-amino-N-ethylanilino)ethanol;sulfuric acid
CCN(CCO)C1=CC=C(C=C1)N.OS(=O)(=O)O
4327-84-8
C10H18N2O5S
278.323
MFCD00040597
KAJALVWKFPQZOO-UHFFFAOYSA-N
N-Ethyl-N-(2-hydroxyethyl)-1,4-phenylenediamine Sulfate
2756529
2-(4-amino-N-ethylanilino)ethanol;sulfuric acid
CCN(CCO)C1=CC=C(C=C1)N.OS(=O)(=O)O
Melting Point 179°C
Color White-Yellow
Quantity 25g
Formula Weight 278.32
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 4-Amino-N-(2-hydroxyethyl)-N-ethylaniline Sulfate

EINECSNumber : (3)-0241&(3)-0246

TSCA : Yes