Alfa Aesar™ 4-Amino-N-Boc-L-phenylalanine, 95%

Manufacturer: Alfa Aesar™ H51980MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51980MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 55533-24-9
    Molecular Formula C14H20N2O4
    Formula Weight 280.32
    MDL Number MFCD00038139
    Synonym Boc-Phe(4-NH2)-OH
    Chemical Name or Material 4-Amino-N-Boc-L-phenylalanine

    Specifications

    Formula Weight 280.32
    Quantity 250mg
    MDL Number MFCD00038139
    Melting Point 124° to 126°C
    Molecular Formula C14H20N2O4
    CAS 55533-24-9
    Synonym Boc-Phe(4-NH2)-OH
    Chemical Name or Material 4-Amino-N-Boc-L-phenylalanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit