Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4′-Aminoacetophenone 98.0+%, TCI America™

Click to view available options
Quantity:
25g
100g
500g
99-92-3
C8H9NO
135.166
MFCD00007896
GPRYKVSEZCQIHD-UHFFFAOYSA-N
4'-aminoacetophenone,4-aminoacetophenone,1-4-aminophenyl ethanone,p-aminoacetophenone,4-acetylaniline,p-acetylaniline,ethanone, 1-4-aminophenyl,p-aminoacetylbenzene,1-4-aminophenyl ethan-1-one,acetophenone, p-amino
7468
1-(4-aminophenyl)ethanone
CC(=O)C1=CC=C(C=C1)N
99-92-3
C8H9NO
135.166
MFCD00007896
GPRYKVSEZCQIHD-UHFFFAOYSA-N
4'-aminoacetophenone,4-aminoacetophenone,1-4-aminophenyl ethanone,p-aminoacetophenone,4-acetylaniline,p-acetylaniline,ethanone, 1-4-aminophenyl,p-aminoacetylbenzene,1-4-aminophenyl ethan-1-one,acetophenone, p-amino
7468
1-(4-aminophenyl)ethanone
CC(=O)C1=CC=C(C=C1)N
Boiling Point 293°C
Color Yellow
Melting Point 107°C
Quantity 500g
Formula Weight 135.17
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 4′-Aminoacetophenone

RTECSNumber : AM5500000

TSCA : Yes