Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminoantipyrine, >98%, Affymetrix/USB™

Click to view available options
Quantity:
50g
100g
Packaging:
Plastic bottle
83-07-8
C11H13N3O
203.245
MFCD00003145
RLFWWDJHLFCNIJ-UHFFFAOYSA-N
4-aminoantipyrine,ampyrone,metapirazone,aminoantipyrine,solvapyrin-a,aminoazophene,4-aminophenazone,4-aminoantipyrene,aminoantipyrin,solnapyrin-a
2151
CHEBI:59026
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N
83-07-8
C11H13N3O
203.245
MFCD00003145
RLFWWDJHLFCNIJ-UHFFFAOYSA-N
4-aminoantipyrine,ampyrone,metapirazone,aminoantipyrine,solvapyrin-a,aminoazophene,4-aminophenazone,4-aminoantipyrene,aminoantipyrin,solnapyrin-a
2151
CHEBI:59026
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N
Packaging Plastic bottle
Quantity 100g
Melting Point 106°C to 110°C
Formula Weight 203.25
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 4-Aminoantipyrine