4-Aminoantipyrine, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

Manufacturer: J.T.Baker A63003

Packaging Wide Mouth Amber Glass Bottle
Quantity 25g

 View more versions of this product

Catalog No. 02-002-105

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 83-07-8
    Molecular Formula C11H13N3O
    Formula Weight 203.24
    Chemical Name or Material 4-Aminoantipyrine

    Specifications

    Formula Weight 203.24
    Molecular Formula C11H13N3O
    CAS 83-07-8
    Packaging Wide Mouth Amber Glass Bottle
    Quantity 25g
    Chemical Name or Material 4-Aminoantipyrine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit