Alfa Aesar™ 4-Aminobenzhydrazide, 98%

Manufacturer: Alfa Aesar™ A1096336

Quantity 500g

 View more versions of this product

Catalog No. AAA1096336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5351-17-7
    Molecular Formula C7H9N3O
    Formula Weight 151.17
    MDL Number MFCD00007606
    Beilstein 639053
    Synonym 4-Aminobenzoic acid hydrazide; (4-Aminobenzoyl)hydrazine
    Chemical Name or Material 4-Aminobenzhydrazide

    Specifications

    Formula Weight 151.17
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 500g
    CAS 5351-17-7
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number DG2580000
    Melting Point 225° to 230°C
    Molecular Formula C7H9N3O
    Beilstein 639053
    MDL Number MFCD00007606
    Synonym 4-Aminobenzoic acid hydrazide; (4-Aminobenzoyl)hydrazine
    Health Hazard 1 Warning
    Chemical Name or Material 4-Aminobenzhydrazide
    Assay 98%

    Description

    Applications
    4-Aminobenzhydrazide has an important role in the peroxidation activity of MPO. It is also used in enzyme preparations and in cell cultures.

    Solubility
    Soluble in DMSO (10 mg/ml), dilute acids, and water (partly miscible).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases. Store at -20°C.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit